MilliporeSigma
PKU-21: A Novel Layered Germanate Built from Ge
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-11-10)
Jie Liang, Jie Su, Fei Peng, Yongfei Zeng, Junliang Sun
ABSTRACT
The attractive properties of layered inorganic materials, which make them suitable for numerous applications in chemical industries and life sciences, originated from their crystalline framework structures. Here, we report a new layered germanate PKU-21, which was prepared by the hydrothermal synthesis method using 2-propanolamine (MIPA) as the structure-directing agent. The structure of PKU-21 was determined from synchrotron single-crystal X-ray diffraction and synchrotron powder X-ray diffraction data. It reveals a complicated framework structure containing 18 unique Ge atoms in the asymmetric unit. PKU-21 is the first layered germanate built from both Ge
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Pricing and availability is not currently available.