MilliporeSigma
  • Proximal Neuropathy and Associated Skeletal Muscle Changes Resembling Denervation Atrophy in Hindlimbs of Chronic Hypoglycaemic Rats.
Proximal Neuropathy and Associated Skeletal Muscle Changes Resembling Denervation Atrophy in Hindlimbs of Chronic Hypoglycaemic Rats.
Basic & clinical pharmacology & toxicology (2017-08-18)
Vivi F H Jensen, Anne-Marie Molck, Henrik Soeborg, Jette Nowak, Melissa Chapman, Jens Lykkesfeldt, Ingrid B Bogh
ABSTRACT
Peripheral neuropathy is one of the most common complications of diabetic hyperglycaemia. Insulin-induced hypoglycaemia (IIH) might potentially exacerbate or contribute to neuropathy as hypoglycaemia also causes peripheral neuropathy. In rats, IIH induces neuropathy associated with skeletal muscle changes. Aims of this study were to investigate the progression and sequence of histopathologic changes caused by chronic IIH in rat peripheral nerves and skeletal muscle, and whether such changes were reversible. Chronic IIH was induced by infusion of human insulin, followed by an infusion-free recovery period in some of the animals. Sciatic, plantar nerves and thigh muscle were examined histopathologically after four or eight weeks of infusion and after the recovery period. IIH resulted in high incidence of axonal degeneration in sciatic nerves and low incidence in plantar nerves indicating proximo-distal progression of the neuropathy. The neuropathy progressed in severity (sciatic nerve) and incidence (sciatic and plantar nerve) with the duration of IIH. The myopathy consisted of groups of angular atrophic myofibres which resembled histopathologic changes classically seen after denervation of skeletal muscle, and severity of the myofibre atrophy correlated with severity of axonal degeneration in sciatic nerve. Both neuropathy and myopathy were still present after four weeks of recovery, although the neuropathy was less severe. In conclusion, the results suggest that peripheral neuropathy induced by IIH progresses proximo-distally, that severity and incidence increase with duration of the hypoglycaemia and that these changes are partially reversible within four weeks. Furthermore, IIH-induced myopathy is most likely secondary to the neuropathy.
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast)−Alkaline Phosphatase antibody produced in mouse, clone MY-32, purified from hybridoma cell culture