MilliporeSigma
  • Synthesis of zeolites with different chemical and textural properties for metal ions removal from aqueous solutions.
Synthesis of zeolites with different chemical and textural properties for metal ions removal from aqueous solutions.
Water science and technology : a journal of the International Association on Water Pollution Research (2017-12-14)
Danielle França de Oliveira, Jefferson Antoniol Mendes Neri, Jonas Alves de Almeida Ribeiro, Fabiana Soares Dos Santos, Mendelssolm Kister de Pietre
ABSTRACT
In this study β-zeolite, ferrierite and partially delaminated PREFER (precursor of ferrierite) zeolites with several chemical and textural properties were synthesized for the removal of zinc and lead ion metals from their respective solutions. Adsorption experiments involving the suspension of tiny amounts of these solids in aqueous solutions containing either Zn
Sigma-Aldrich
4-Amino-2,2,6,6-tetramethylpiperidine, 98%
Pricing and availability is not currently available.