MilliporeSigma
  • Sophoridine suppresses cell growth in human medulloblastoma through FoxM1, NF-κB and AP-1.
Sophoridine suppresses cell growth in human medulloblastoma through FoxM1, NF-κB and AP-1.
Oncology letters (2018-01-19)
Zhensong Yue, Tongguo Si, Zhanyu Pan, Wenfeng Cao, Zhuchen Yan, Zhansheng Jiang, Huaqiang Ouyang
ABSTRACT
Sophoridine is an alkaloid extracted from
Sigma-Aldrich
N-Acetyl-Ile-Glu-Thr-Asp-p-nitroanilide, ≥95%
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, ≥97% (HPLC), powder