MilliporeSigma
Icaritin inhibits decidualization of endometrial stromal cells.
Experimental and therapeutic medicine (2017-12-30)
Aiwen Le, Zhong Hai Wang, Xiao Yun Dai, Tian Hui Xiao, Rong Zhuo, Baozhen Zhang, Zhonglin Xiao, Xiujun Fan
ABSTRACT
The aim of the present study was to investigate the effects of Icaritin on the proliferation and decidualization of endometrial stromal cells (ESCs). A total of 20 specimens of endometrium were collected during hysterectomy at the Gynecology Department of Shenzhen Nanshan People's Hospital (Shenzhen, China) between August 2014 and December 2015. The endometrium was digested with high concentrations of collagenase and DNase and filtered with meshes, and then the glandular epithelial and stromal cells were separated by the adhesion purification method. The purity of stromal cells was identified by vimetin and cytokeratin 7 immunostaining. The estradiol + progesterone (E2+P4) and/or cyclic adenosine monophosphate (cAMP) were added to induce an
Sigma-Aldrich
Icaritin, ≥98% (HPLC)
Sigma-Aldrich
Human IGFBP-1 ELISA Kit, for serum, plasma, cell culture supernatant and urine