MilliporeSigma
  • Leucine supplementation after mechanical stimulation activates protein synthesis via L-type amino acid transporter 1 in vitro.
Leucine supplementation after mechanical stimulation activates protein synthesis via L-type amino acid transporter 1 in vitro.
Journal of cellular biochemistry (2017-09-01)
Naoya Nakai, Fuminori Kawano, Taro Murakami, Ken Nakata, Kazuhiko Higashida
ABSTRACT
Branched-chain amino acid supplements consumed following exercise are widely used to increase muscle mass. Although both exercise (ie, mechanical stimulation) and branched-chain amino acid leucine supplementation have been reported to stimulate muscle protein synthesis by activating the mammalian target of rapamycin (mTOR) signaling pathway independently, the mechanisms underlying their synergistic effects are largely unknown. Utilizing cultured differentiated C2C12 myotubes, we established a combination treatment model in which the cells were subjected to cyclic uniaxial mechanical stretching (4 h, 15%, 1 Hz) followed by stimulation with 2 mM leucine for 45 min. Phosphorylation of p70 S6 kinase (p70S6K), an mTOR-regulated marker of protein translation initiation, was significantly increased following mechanical stretching alone but returned to the baseline after 4 h. Leucine supplementation further increased p70S6K phosphorylation, with a greater increase observed in the stretched cells than in the non-stretched cells. Notably, the expression of L-type amino acid transporter 1 (LAT1), a stimulator of the mTOR pathway, was also increased by mechanical stretching, and siRNA-mediated knockdown partially attenuated leucine-induced p70S6K phosphorylation. These results suggest that mechanical stretching promotes LAT1 expression and, consequently, amino acid uptake, leading to enhanced leucine-induced activation of protein synthesis. LAT1 has been demonstrated to be a point of crosstalk between exercise- and nutrition-induced skeletal muscle growth.
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC7A5