MilliporeSigma
Establishment and characterization of the Bombyx mandarina cell line.
Journal of invertebrate pathology (2009-05-23)
Masashi Iwanaga, Rika Arai, Yuka Shibano, Hideki Kawasaki, Shigeo Imanishi
ABSTRACT
A new cell line, designated as NIAS-Boma-529b, was established from the larval fat bodies of Bombyx mandarina (B. mandarina), which is believed to be an ancestor of Bombyx mori (B. mori). This cell line has been cultured for approximately 150 passages during 2years in an IPL-41 medium supplemented with 10% fetal bovine serum at a constant temperature of 26 degrees C. The morphology of this line includes adhesive round and spindle-shaped cells. Random-amplified polymorphic DNA analysis (RAPD) using 7 primers and a statistical analysis based on Nei's genetic distance revealed that this cell line was closely related to B. mori-derived cell lines. An infection study also revealed that this cell line was susceptible to B. mori nucleopolyhedrovirus (BmNPV); however, it had no apparent susceptibility to Autographa californica NPV (AcNPV), which is closely related to BmNPV. Nevertheless, cells infected with AcNPV showed an extensive cytopathic effect (CPE), including a rough cell surface, rounding, nuclear expansion, and cell blebbing. These results suggest that this cell line can be useful to clarify the mechanism of host range determination of BmNPV and AcNPV.
Sigma-Aldrich
IPL-41 Insect Medium, With L-glutamine and sodium bicarbonate., liquid, sterile-filtered, suitable for insect cell culture