MilliporeSigma
  • Uncoupling protein-1 is protective of bone mass under mild cold stress conditions.
Uncoupling protein-1 is protective of bone mass under mild cold stress conditions.
Bone (2015-06-10)
Amy D Nguyen, Nicola J Lee, Natalie K Y Wee, Lei Zhang, Ronaldo F Enriquez, Ee Cheng Khor, Tao Nie, Donghai Wu, Amanda Sainsbury, Paul A Baldock, Herbert Herzog
ABSTRACT
Brown adipose tissue (BAT), largely controlled by the sympathetic nervous system (SNS), has the ability to dissipate energy in the form of heat through the actions of uncoupling protein-1 (UCP-1), thereby critically influencing energy expenditure. Besides BAT, the SNS also strongly influences bone, and recent studies have demonstrated a positive correlation between BAT activity and bone mass, albeit the interactions between BAT and bone remain unclear. Here we show that UCP-1 is critical for protecting bone mass in mice under conditions of permanent mild cold stress for this species (22°C). UCP-1
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.