MilliporeSigma
  • Parathyroid Hormone-Related Protein Promotes Rat Stem Leydig Cell Differentiation.
Parathyroid Hormone-Related Protein Promotes Rat Stem Leydig Cell Differentiation.
Frontiers in physiology (2017-11-29)
Tiantian Song, Yiyan Wang, Huitao Li, Lanlan Chen, Jianpeng Liu, Xianwu Chen, Xiaojun Li, Xiaoheng Li, Linxi Li, Qingquan Lian, Ren-Shan Ge
ABSTRACT
The regulatory factors for stem Leydig cell development are largely unknown. Herein, we reported that parathyroid hormone-related protein (PTHrP) may be a factor to regulate this process. The effects of PTHrP on rat stem Leydig cell proliferation and differentiation were investigated using a stem Leydig cell culture system and an ethane dimethane sulfonate (EDS)-treated
Sigma-Aldrich
Human LHB / Lutropin Subunit Beta ELISA Kit