MilliporeSigma
  • Analysis of the respiratory component of heart rate variability in the Cururu toad Rhinella schneideri.
Analysis of the respiratory component of heart rate variability in the Cururu toad Rhinella schneideri.
Scientific reports (2017-11-25)
Lucas A Zena, Cléo A C Leite, Leonardo S Longhini, Daniel P M Dias, Glauber S F da Silva, Lynn K Hartzler, Luciane H Gargaglioni, Kênia C Bícego
ABSTRACT
Beat-to-beat variation in heart rate (f
Sigma-Aldrich
Ethyl 3-aminobenzoate, 97%