- Sucrosomial® iron absorption studied by in vitro and ex-vivo models.
Sucrosomial® iron absorption studied by in vitro and ex-vivo models.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2017-10-23)
Angela Fabiano, Elisa Brilli, Stefano Fogli, Denise Beconcini, Sara Carpi, Germano Tarantino, Ylenia Zambito
PMID29055735
ABSTRACT
This paper presents a comparative evaluation of different oral ferric iron formulations for ability to retain Fe