MilliporeSigma
  • Regulation of TG accumulation and lipid droplet morphology by the novel TLDP1 in
Regulation of TG accumulation and lipid droplet morphology by the novel TLDP1 in
Journal of lipid research (2017-10-14)
Takashi Watanabe, Ryo Sakiyama, Yuya Iimi, Satomi Sekine, Eriko Abe, Kazuko H Nomura, Kazuya Nomura, Yohei Ishibashi, Nozomu Okino, Masahiro Hayashi, Makoto Ito
ABSTRACT
Thraustochytrids are marine single-cell protists that produce large amounts of PUFAs, such as DHA. They accumulate PUFAs in lipid droplets (LDs), mainly as constituent(s) of triacylglycerol (TG). We identified a novel protein in the LD fraction of
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycerol, ≥99%