MilliporeSigma
Influence of Porosity and Pore-Size Distribution in Ti
ACS applied materials & interfaces (2017-10-24)
Kausik Kapat, Pavan Kumar Srivas, Arun Prabhu Rameshbabu, Priti Prasanna Maity, Subhodeep Jana, Joy Dutta, Pallab Majumdar, Debalay Chakrabarti, Santanu Dhara
ABSTRACT
Cementless fixation for orthopedic implants aims to obviate challenges associated with bone cement, providing long-term stability of bone prostheses after implantation. The application of porous titanium and its alloy-based implants is emerging for load-bearing applications due to their high specific strength, low stiffness, corrosion resistance, and superior osteoconductivity. In this study, coagulant-assisted foaming was utilized for the fabrication of porous Ti
Sigma-Aldrich
DNA Quantitation Kit, Fluorescence Assay, Quantitation of DNA using bisBenzimide