MilliporeSigma
  • EZH2 promotes degradation of stalled replication forks by recruiting MUS81 through histone H3 trimethylation.
EZH2 promotes degradation of stalled replication forks by recruiting MUS81 through histone H3 trimethylation.
Nature cell biology (2017-10-17)
Beatrice Rondinelli, Ewa Gogola, Hatice Yücel, Alexandra A Duarte, Marieke van de Ven, Roxanne van der Sluijs, Panagiotis A Konstantinopoulos, Jos Jonkers, Raphaël Ceccaldi, Sven Rottenberg, Alan D D'Andrea
ABSTRACT
The emergence of resistance to poly-ADP-ribose polymerase inhibitors (PARPi) poses a threat to the treatment of BRCA1 and BRCA2 (BRCA1/2)-deficient tumours. Stabilization of stalled DNA replication forks is a recently identified PARPi-resistance mechanism that promotes genomic stability in BRCA1/2-deficient cancers. Dissecting the molecular pathways controlling genomic stability at stalled forks is critical. Here we show that EZH2 localizes at stalled forks where it methylates Lys27 on histone 3 (H3K27me3), mediating recruitment of the MUS81 nuclease. Low EZH2 levels reduce H3K27 methylation, prevent MUS81 recruitment at stalled forks and cause fork stabilization. As a consequence, loss of function of the EZH2/MUS81 axis promotes PARPi resistance in BRCA2-deficient cells. Accordingly, low EZH2 or MUS81 expression levels predict chemoresistance and poor outcome in patients with BRCA2-mutated tumours. Moreover, inhibition of Ezh2 in a murine Brca2
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Pricing and availability is not currently available.
Sigma-Aldrich
5-Chloro-2′-deoxyuridine, thymidine analog
Pricing and availability is not currently available.