MilliporeSigma
  • A model to describe the inhomogeneous broadening of NQR lines in chlorohalobenzenes with orientational or substitutional disorder.
A model to describe the inhomogeneous broadening of NQR lines in chlorohalobenzenes with orientational or substitutional disorder.
Solid state nuclear magnetic resonance (2017-07-01)
Silvina C Pérez, Mariano Zuriaga
ABSTRACT
A simple model to explain the NQR lineshape in solids with orientational disorder or substitutional disorder is presented. The particular case of m-chlorobromobenzene is studied. It is based on the assumption that Bromine atoms, of m-chlorobromobenzene molecules, behave as point defects in the m-dichlorobenzene lattice that modify the crystalline Electric Field Gradient. The model is also tested successfully in solid solutions of p-dichlorobenzene-p-dibromobenzene, where Bromine atoms of p-dibromobenzene molecules are assumed to be homogeneously distributed in the p-dichlorobenzene lattice. The lineshape, of others disordered chlorohalobenzenes, are also analyzed. Also, a characterization of m-chlorobromobenzene dynamics is included. In particular, there is no evidence of molecular reorientations as it is observed in the disordered phases of o-chlorobromobenzene.
Sigma-Aldrich
1-Bromo-3-chlorobenzene, 99%