MilliporeSigma
  • Poly(oligoethylene glycol methacrylate) dip-coating: turning cellulose paper into a protein-repellent platform for biosensors.
Poly(oligoethylene glycol methacrylate) dip-coating: turning cellulose paper into a protein-repellent platform for biosensors.
Journal of the American Chemical Society (2014-08-30)
Xudong Deng, Niels M B Smeets, Clémence Sicard, Jingyun Wang, John D Brennan, Carlos D M Filipe, Todd Hoare
ABSTRACT
The passivation of nonspecific protein adsorption to paper is a major barrier to the use of paper as a platform for microfluidic bioassays. Herein we describe a simple, scalable protocol based on adsorption and cross-linking of poly(oligoethylene glycol methacrylate) (POEGMA) derivatives that reduces nonspecific adsorption of a range of proteins to filter paper by at least 1 order of magnitude without significantly changing the fiber morphology or paper macroporosity. A lateral-flow test strip coated with POEGMA facilitates effective protein transport while also confining the colorimetric reporting signal for easier detection, giving improved performance relative to bovine serum albumin (BSA)-blocked paper. Enzyme-linked immunosorbent assays based on POEGMA-coated paper also achieve lower blank values, higher sensitivities, and lower detection limits relative to ones based on paper blocked with BSA or skim milk. We anticipate that POEGMA-coated paper can function as a platform for the design of portable, disposable, and low-cost paper-based biosensors.
Sigma-Aldrich
Poly(M(EO)2MA:Poly(OEGMA), (90:10), aldehyde functionalized, 25 wt% solution in water
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(OEGMA), hydrazide functionalized, 25 wt% solution in water
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(M(EO)2MA:Poly(OEGMA) 90:10, hydrazide functionalized, 25 wt. % (solution in water)
Pricing and availability is not currently available.