MilliporeSigma
  • Community structure follows simple assembly rules in microbial microcosms.
Community structure follows simple assembly rules in microbial microcosms.
Nature ecology & evolution (2017-08-16)
Jonathan Friedman, Logan M Higgins, Jeff Gore
ABSTRACT
Microorganisms typically form diverse communities of interacting species, whose activities have tremendous impact on the plants, animals and humans they associate with. The ability to predict the structure of these complex communities is crucial to understanding and managing them. Here, we propose a simple, qualitative assembly rule that predicts community structure from the outcomes of competitions between small sets of species, and experimentally assess its predictive power using synthetic microbial communities composed of up to eight soil bacterial species. Nearly all competitions resulted in a unique, stable community, whose composition was independent of the initial species fractions. Survival in three-species competitions was predicted by the pairwise outcomes with an accuracy of ~90%. Obtaining a similar level of accuracy in competitions between sets of seven or all eight species required incorporating additional information regarding the outcomes of the three-species competitions. Our results demonstrate experimentally the ability of a simple bottom-up approach to predict community structure. Such an approach is key for anticipating the response of communities to changing environments, designing interventions to steer existing communities to more desirable states and, ultimately, rationally designing communities de novo.
Sigma-Aldrich
M9, Minimal Salts, 5X, powder, minimal microbial growth medium
Sigma-Aldrich
Thyroglobulin from bovine thyroid, powder, ≥90% (agarose gel electrophoresis)