MilliporeSigma
  • The anti-proliferative effects of oleanolic acid on A7r5 cells-Role of UCP2 and downstream FGF-2/p53/TSP-1.
The anti-proliferative effects of oleanolic acid on A7r5 cells-Role of UCP2 and downstream FGF-2/p53/TSP-1.
Cell biology international (2017-08-10)
Yantao Han, Qixiao Jiang, Yu Wang, Wenqian Li, Min Geng, Zhiwu Han, Xuehong Chen
ABSTRACT
Vascular smooth muscle cell (VSMC) proliferation is a major contributor to atherosclerosis. This study investigated the inhibitory effects of oleanolic acid (OA) against oxidized low-density lipoprotein (ox-LDL)-induced VSMC proliferation in A7r5 cells and explored underlying molecular mechanism. The cell proliferation was quantified with cell counting kit-8 (CCK-8), in which ox-LDL significantly increased A7r5 cells proliferation, while OA pretreatment effectively alleviated such changes without inducing overt cytotoxicity, as indicated by lactate dehydrogenase (LDH) assay. Quantitative real-time RT-PCR (qRT-PCR) and Western blotting revealed increased UCP2 and FGF-2 expression levels as well as decreased p53 and TSP-1 expression levels in A7r5 cells following ox-LDL exposure, while OA pretreatment reversed such changes. Furthermore, inhibiting UCP2 with genipin remarkably reversed the changes in the expression levels of FGF-2, p53, and TSP-1 induced by ox-LDL exposure; silencing FGF-2 with siRNA did not significantly change the expression levels of UCP2 but effectively reversed the changes in the expression levels of p53 and TSP-1, and activation of p53 with PRIMA-1 only significantly affected the changes in the expression levels of TSP-1, but not in UCP2 or FGF-2, suggesting a UCP-2/FGF-2/p53/TSP-1 signaling in A7r5 cells response to ox-LDL exposure. Additionally, co-treatment of OA and genipin exhibited similar effects to the expression levels of UCP2, FGF-2, p53, and TSP-1 as OA or genipin solo treatment in ox-LDL-exposed A7r5 cells, suggesting the involvement of UCP-2/FGF-2/p53/TSP-1 in the mechanism of OA. In conclusion, OA inhibits ox-LDL-induced VSMC proliferation in A7r5 cells, the mechanism involves the changes in UCP-2/FGF-2/p53/TSP-1.
Sigma-Aldrich
PRIMA-1, ≥98% (HPLC), solid
Sigma-Aldrich
MISSION® esiRNA, targeting human FGF2