MilliporeSigma
Reduction of 4(5)-Methylimidazole Using Cookie Model Systems.
Journal of food science (2017-09-12)
Min-Chul Jung, Mina K Kim, Kwang-Geun Lee
ABSTRACT
The objective of this study was to determine the reduction of 4(5)-methylimidazole (4-MI) under various baking conditions. For 4-MI analysis, an analytical method using gas chromatography-mass spectrometry was developed. The developed method was validated with linearity (r A consumer awareness toward toxicity of 4-MI has been arising, and method to reduce the levels of 4-MI in food products are being developed in many studies. Yet, these reduction studies in food model systems only focused on use of food additives for 4-MI reduction. Current study investigated the use of process modification on 4-MI reduction in cookie, and suggested that baking cookies longer at lower temperature, in turn, reduces the levels of 4-MI in cookies without compromising consumer acceptance. Finding from current study can practically aid bakery industry to ensure safety of bakery products without affecting consumer likings.
Sigma-Aldrich
2-Ethylimidazole, 98%