MilliporeSigma
  • Multifunctional nanocomposite hollow fiber membranes by solvent transfer induced phase separation.
Multifunctional nanocomposite hollow fiber membranes by solvent transfer induced phase separation.
Nature communications (2017-11-02)
Martin F Haase, Harim Jeon, Noah Hough, Jong Hak Kim, Kathleen J Stebe, Daeyeon Lee
ABSTRACT
The decoration of porous membranes with a dense layer of nanoparticles imparts useful functionality and can enhance membrane separation and anti-fouling properties. However, manufacturing of nanoparticle-coated membranes requires multiple steps and tedious processing. Here, we introduce a facile single-step method in which bicontinuous interfacially jammed emulsions are used to form nanoparticle-functionalized hollow fiber membranes. The resulting nanocomposite membranes prepared via solvent transfer-induced phase separation and photopolymerization have exceptionally high nanoparticle loadings (up to 50 wt% silica nanoparticles) and feature densely packed nanoparticles uniformly distributed over the entire membrane surfaces. These structurally well-defined, asymmetric membranes facilitate control over membrane flux and selectivity, enable the formation of stimuli responsive hydrogel nanocomposite membranes, and can be easily modified to introduce antifouling features. This approach forms a foundation for the formation of advanced nanocomposite membranes comprising diverse building blocks with potential applications in water treatment, industrial separations and as catalytic membrane reactors.
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%