MilliporeSigma
  • Firefly-mimicking intensive and long-lasting chemiluminescence hydrogels.
Firefly-mimicking intensive and long-lasting chemiluminescence hydrogels.
Nature communications (2017-10-19)
Yating Liu, Wen Shen, Qi Li, Jiangnan Shu, Lingfeng Gao, Mingming Ma, Wei Wang, Hua Cui
ABSTRACT
Most known chemiluminescence (CL) reactions exhibit flash-type light emission. Great efforts have been devoted to the development of CL systems that emit light with high intensity and long-lasting time. However, a long-lasting CL system that can last for hundreds of hours is yet-to-be-demonstrated. Here we show firefly-mimicking intensive and long-lasting CL hydrogels consisting of chitosan, CL reagent N-(4-aminobutyl)-N-ethylisoluminol (ABEI) and catalyst Co
Sigma-Aldrich
N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%