MilliporeSigma
  • Structural and serological studies on a new acidic O-specific polysaccharide of Proteus vulgaris O32.
Structural and serological studies on a new acidic O-specific polysaccharide of Proteus vulgaris O32.
European journal of biochemistry (1998-10-06)
B Bartodziejska, A S Shashkov, D Babicka, A A Grachev, A Torzewska, N A Paramonov, A Y Chernyak, A Rozalski, Y A Knirel
ABSTRACT
The following structure of the O-specific polysaccharide chain (O-antigen) of the Proteus vulgaris 032 lipopolysaccharide (LPS) was established by 1H-NMR and 13C-NMR spectroscopy, including two-dimensional NOESY and H-detected 1H,13C heteronuclear multiple-quantum coherence (HMQC) experiments: -->2)-alpha-L-RhapI-(1-->2)-alpha-L-RhapII-(1-->4)-beta-D-++ +GalpA(I)-(1-->3)-beta-D-GlcpNAc-(1-->4)-alpha-D-GalpA(II)-(1-- >. In addition, an O-acetyl group was detected, which, most probably, is located at position 3 of a part of RhapI residues. Serological studies, using rabbit polyclonal anti-(P. vulgaris 032) serum, homologous and heterologous Proteus O-antigens and related artificial antigens, revealed the importance of an a-D-GalA-associated epitope in manifesting the immunospecificity of P. vulgaris 032 and substantiated serological relationships between the O-antigen studied and those of some other Proteus strains.
Sigma-Aldrich
Lipopolysaccharides from Proteus vulgaris, purified by phenol extraction