MilliporeSigma
  • Ferulic acid promotes osteogenesis of bone marrow-derived mesenchymal stem cells by inhibiting microRNA-340 to induce β-catenin expression through hypoxia.
Ferulic acid promotes osteogenesis of bone marrow-derived mesenchymal stem cells by inhibiting microRNA-340 to induce β-catenin expression through hypoxia.
European journal of cell biology (2017-08-03)
Kewei Du, Ziqiang Li, Xuchen Fang, Tingwei Cao, Yaozeng Xu
ABSTRACT
Osteogenic differentiation is regulated through multiple signaling networks that may include responses to hypoxia. Antioxidant ferulic acid (FA) can promote hypoxia signaling by inducing hypoxic-induced factor (HIF). However, whether FA could affect osteogenesis has not been explored. We examined human bone marrow-derived mesenchymal stem cell (MSC) following FA treatment. The expression of β-catenin was measured, and candidate microRNAs that target β-catenin were studied. The involvement of hypoxia was investigated in miR-340-5p that contains hypoxia response elements (HRE) in the promoter region. Further, the osteogenic potential of FA-treated MSC was assessed by alkaline phosphatase (ALP) activity and alizarin red staining assays. Osteoblast marker gene expressions were also compared between controls and FA-treated cells. FA induced β-catenin expression in MSC. This effect is likely mediated through a derepression of β-catenin 3'-UTR inhibition by miR-340-5p. HIF-1α, which suppressed miR-340-5p promoter activation through HRE motifs, was induced by FA. The induction of β-catenin signaling by FA was consistent with an enhancement in osteogenesis of FA-treated MSC, which could be attenuated by miR-340-5p overexpression. Analysis of the signaling networks induced by FA reveals that hypoxia may promote the osteogenic program in mesenchymal stem cells via a novel microRNA pathway.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)