MilliporeSigma
Unravelling redox processes of Li
Physical chemistry chemical physics : PCCP (2017-10-04)
D Muller-Bouvet, N Emery, N Tassali, E Panabière, S Bach, O Crosnier, T Brousse, C Cénac-Morthe, A Michalowicz, J P Pereira-Ramos
ABSTRACT
A large data set of XAS (X-ray Absorption Spectroscopy) Manganese K-edge spectra has been collected operando and studied upon the electrochemical oxidation of the promising Li-ion battery anode material Li
Sigma-Aldrich
Lithium nitride, −60 mesh, ≥99.5%
Pricing and availability is not currently available.