MilliporeSigma
  • Low antigen dose formulated in CAF09 adjuvant Favours a cytotoxic T-cell response following intraperitoneal immunization in Göttingen minipigs.
Low antigen dose formulated in CAF09 adjuvant Favours a cytotoxic T-cell response following intraperitoneal immunization in Göttingen minipigs.
Vaccine (2017-09-10)
Nana H Overgaard, Thomas M Frøsig, Jeanne T Jakobsen, Søren Buus, Mads H Andersen, Gregers Jungersen
ABSTRACT
The relationship between the antigen dose and the quality of an immune response generated upon immunization is poorly understood. However, findings show that the immune system is indeed influenced by the antigen dose; hence underlining the importance of correctly determining which dose to use in order to generate a certain type of immune response. To investigate this area further, we used Göttingen minipigs asan animal model especially due to the similar body size and high degree of immunome similarity between humans and pigs. In this study, we show that both a humoral and a cell-mediated immune (CMI) response can be generated following intraperitoneal immunization with tetanus toxoid (TT) formulated in the CAF09 liposomal adjuvant. Importantly, a low antigen dose induced more TT-specific polyfunctional T cells, whereas antigen-specific IgG production was observed upon high-dose immunization. Independent of antigen dose, intraperitoneal administration of antigen increased the amount of TT-specific cytotoxic CD8β
Sigma-Aldrich
BCIP®/NBT Liquid Substrate System, ready to use solution
Pricing and availability is not currently available.