MilliporeSigma
  • Detoxification of diphenyl ether herbicide lactofen by Bacillus sp. Za and enantioselective characteristics of an esterase gene lacE.
Detoxification of diphenyl ether herbicide lactofen by Bacillus sp. Za and enantioselective characteristics of an esterase gene lacE.
Journal of hazardous materials (2017-08-13)
Jing Zhang, Luyao Lu, Feng Chen, Lingling Chen, Jingang Yin, Xing Huang
ABSTRACT
A bacterial strain Za capable of degrading diphenyl ether herbicide lactofen was isolated and identified as Bacillus sp. This strain could degrade 94.8% of 50mgL
Product Description
Supelco
Lactofen, PESTANAL®, analytical standard