MilliporeSigma
  • Quaternary Ammonium Polyamidoamine Dendrimer Modified Quantum Dots as Fluorescent Probes for p-Fluorophenoxyacetic Acid Detection in Aqueous Solution.
Quaternary Ammonium Polyamidoamine Dendrimer Modified Quantum Dots as Fluorescent Probes for p-Fluorophenoxyacetic Acid Detection in Aqueous Solution.
Journal of fluorescence (2017-09-13)
Xiao Xu, Yuan-Cheng Cao, Jun'An Liu, Yongjun Lin
ABSTRACT
The wide use of pesticide p-fluorophenoxyacetic acid has caused the serious environmental contaminant. A novel fluorescent probe for sensitive detection of p-fluorophenoxyacetic acid in aqueous solutions based on 3.0G quaternary ammonium polyamidoamine (PAMAM) dendrimer modified quantum dots (QDs) (PAMAM@QDs) was reported. Through the solvent-evaporation method, quaternary ammonium PAMAM was employed to modify the QDs. Poloxamer 188 was used to improve the solubility and stability. The resultant PAMAM@QDs dispersed well in water. Fluorescence (FL) spectroscopic study showed that the FL intensity of the PAMAM@QDs was enhanced in the presence of p-fluorophenoxyacetic acid. Under optimal conditions, the enhanced FL intensity as a function of concentration matched very well in the range of 1 ~ 200 µg/mL of p-fluorophenoxyacetic acid, while the lower limits of detection were found to be 0.16 µg/mL. These results show that PAMAM@QDs is a promising luminescent probe for the detection of pesticides.