MilliporeSigma
  • Clotrimazole microemulsion and microemulsion-based gel: evaluation of buccal drug delivery and irritancy using chick chorioallantoic membrane as the model.
Clotrimazole microemulsion and microemulsion-based gel: evaluation of buccal drug delivery and irritancy using chick chorioallantoic membrane as the model.
The Journal of pharmacy and pharmacology (2017-08-25)
Jarika Kaewbanjong, Paul Wan Sia Heng, Prapaporn Boonme
ABSTRACT
To investigate the efficacy of clotrimazole microemulsion (CTZ-ME) and its gel form, clotrimazole microemulsion-based gel (CTZ-MBG), for the treatment of oral candidiasis. CTZ-ME and CTZ-MBG were characterized for droplet size and texture, respectively. The ex-vivo permeation study and irritancy assessment of CTZ-ME and CTZ-MBG were performed using chick chorioallantoic membrane (CAM) as the model. Antifungal activity against Candida albicans ATCC 10 231 of CTZ-ME and CTZ-MBG was determined by agar diffusion method compared to the blank counterparts. CTZ-ME contained nano-sized droplets and CTZ-MBG had acceptable firmness and spreadability. CTZ-ME exhibited faster CAM permeation of the drug and larger inhibition zone than CTZ-MBG as the increased viscosity of CTZ-MBG resulted in more retardation and higher fluctuations in drug diffusion. As there were no detectable visual changes in CAM blood vessels after applying CTZ-ME or CTZ-MBG, both formulations were non-irritants. CTZ-ME and CTZ-MBG could deliver the drug through CAM, the model for buccal delivery. Additionally, they did not cause irritancy and had effective antifungal activity against C. albicans. The results indicated that CTZ-ME and CTZ-MBG were potential effective antifungal formulations to treat oral candidiasis.
Sigma-Aldrich
Isopentenyl pyrophosphate triammonium salt solution, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)