MilliporeSigma
  • Comparative Analysis of Chrysanthemum Stunt Viroid Accumulation and Movement in Two Chrysanthemum (
Comparative Analysis of Chrysanthemum Stunt Viroid Accumulation and Movement in Two Chrysanthemum (
Frontiers in plant science (2017-12-19)
Tomoyuki Nabeshima, Motoaki Doi, Munetaka Hosokawa
ABSTRACT
Chrysanthemum stunt viroid (CSVd) was inoculated into two chrysanthemum (
Sigma-Aldrich
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%