MilliporeSigma
  • How does oestradiol influence the AVT/IT system in female round gobies during different reproductive phases?
How does oestradiol influence the AVT/IT system in female round gobies during different reproductive phases?
Biology open (2017-09-02)
Hanna Kalamarz-Kubiak, Magdalena Gozdowska, Tatiana Guellard, Ewa Kulczykowska
ABSTRACT
In this
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
Mayer′s Hematoxylin Solution