MilliporeSigma
  • The adsorption of α-amylase on barley proteins affects the in vitro digestion of starch in barley flour.
The adsorption of α-amylase on barley proteins affects the in vitro digestion of starch in barley flour.
Food chemistry (2017-09-30)
Wenwen Yu, Wei Zou, Sushil Dhital, Peng Wu, Michael J Gidley, Glen P Fox, Robert G Gilbert
ABSTRACT
The conversion of barley starch to sugars is a complex enzymic process. Most previous work concerned the biotechnical aspect of in situ barley enzymes. However, the interactions among the macromolecular substrates and their effects on enzymic catalysis has been little examined. Here, we explore the mechanisms whereby interactions of protein and starch in barley flour affect the kinetics of enzymatic hydrolysis of starch in an in vitro system, using digestion rate data and structural analysis by confocal microscopy. The degradation kinetics of both uncooked barley flour and of purified starches are found to be two-step sequential processes. Barley proteins, especially the water-soluble component, are found to retard the digestion of starch degraded by α-amylase: the enzyme binds with water-insoluble protein and with starch granules, leading to reduced starch hydrolysis. These findings are of potential industrial value in both the brewing and food industries.
Sigma-Aldrich
Phosphate buffered saline, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Pricing and availability is not currently available.