MilliporeSigma
Origin of Fine Particulate Carbon in the Rural United States.
Environmental science & technology (2017-08-02)
Bret A Schichtel, Jenny L Hand, Michael G Barna, Kristi A Gebhart, Scott Copeland, John Vimont, William C Malm
ABSTRACT
Carbonaceous compounds are a significant component of fine particulate matter and haze in national parks and wilderness areas where visibility is protected, i.e., class I areas (CIAs). The Regional Haze Rule set the goal of returning visibility in CIAs on the most anthropogenically impaired days to natural by 2064. To achieve this goal, we need to understand contributions of natural and anthropogenic sources to the total fine particulate carbon (TC). A Lagrangian chemical transport model was used to simulate the 2006-2008 contributions from various source types to measured TC in CIAs and other rural lands. These initial results were incorporated into a hybrid model to reduce systematic biases. During summer months, fires and vegetation-derived secondary organic carbon together often accounted for >75% of TC. Smaller contributions, <20%, from area and mobile sources also occurred. During the winter, contributions from area and mobile sources increased, with area sources accounting for half or more of the TC in many regions. The area emissions were likely primarily from residential and industrial wood combustion. Different fire seasons were evident, with the largest contributions during the summer when wildfires occur and smaller contributions during the spring and fall when prescribed and agricultural fires regularly occur.
Sigma-Aldrich
1,6-Anhydro-β-D-glucose, 99%