MilliporeSigma
Elongation of very Long-Chain (>C
Lipids (2017-09-01)
Angela Oboh, Juan C Navarro, Douglas R Tocher, Oscar Monroig
ABSTRACT
Elongation of very long-chain fatty acid 4 (Elovl4) proteins participate in the biosynthesis of very long-chain (>C
Sigma-Aldrich
Stearidonic acid, ≥99%