MilliporeSigma
EP4 receptor promotes invadopodia and invasion in human breast cancer.
European journal of cell biology (2017-01-18)
Felix Tönisen, Louisiane Perrin, Battuya Bayarmagnai, Koen van den Dries, Alessandra Cambi, Bojana Gligorijevic
ABSTRACT
The production of Prostaglandin E
Sigma-Aldrich
MISSION® esiRNA, targeting human PTGER4