MilliporeSigma
Protein
Journal of biomolecular NMR (2017-08-02)
Yoshikazu Hattori, David Heidenreich, Yuki Ono, Toshihiko Sugiki, Kei-Ichi Yokoyama, Ei-Ichiro Suzuki, Toshimichi Fujiwara, Chojiro Kojima
ABSTRACT
The preparation of stable isotope-labeled proteins is important for NMR studies, however, it is often hampered in the case of eukaryotic proteins which are not readily expressed in Escherichia coli. Such proteins are often conveniently investigated following post-expression chemical isotope tagging. Enzymatic
Sigma-Aldrich
2,2,2-Trifluoroethylamine hydrochloride, 98%