MilliporeSigma
  • Effects of photoperiod modulation and melatonin feeding around drying-off on bovine mammary gland involution.
Effects of photoperiod modulation and melatonin feeding around drying-off on bovine mammary gland involution.
Journal of dairy science (2017-08-02)
B Ponchon, P Lacasse, S Ollier, X Zhao
ABSTRACT
The risk for a dairy cow to acquire new intramammary infections is high during the transition from lactation to the dry period, because of udder engorgement and altered immune functions. Once the gland is fully involuted, it becomes much more resistant to intramammary infections. Therefore, strategies to depress milk yield before drying-off and accelerate the involution process after drying-off could be beneficial for udder health. The objective of this study was to assess the effect of photoperiod manipulation and melatonin feeding from 14 d before to 14 d after drying-off on the speed of the involution process. Thirty Holstein cows in late lactation were randomly allocated to one of the following treatments: (1) a long-day photoperiod (16 h of light: 8 h of darkness), (2) a short-day photoperiod (8 h of light: 16 h of darkness), and (3) a long-day photoperiod supplemented by melatonin feeding (4 mg/kg of body weight). Milk and blood samples were collected on d -26, -19, -12, -5, -1, 1, 3, 5, 7, 10, and 14 relative to the last milking to determine concentrations of mammary gland involution markers and serum prolactin. Additional blood samples were taken around milking on d -15, before the start of the treatments, and on d -1, before drying-off, to evaluate the treatment effects on milking-induced prolactin release. The short-day photoperiod slightly decreased milk production and basal prolactin secretion during the dry period. The milking-induced prolactin surge was smaller on d -1 than on d -15 regardless of the treatments. Lactoferrin concentration, somatic cell count, and BSA concentration as well as matrix metalloproteinase-2 and -9 activities increased in mammary secretions during the first 2 wk of the dry period, whereas milk citrate concentration and the citrate:lactoferrin molar ratio decreased. The rates of change of these parameters were not significantly affected by the treatments. The long-day photoperiod supplemented by melatonin feeding did not affect milk production, prolactin secretion, or mammary gland involution. Under the conditions in this study, photoperiod modulation and melatonin feeding did not appear to affect the rate of mammary gland involution.
Sigma-Aldrich
Human LTF / Lactotransferrin ELISA Kit