MilliporeSigma
Genome-wide identification of bacterial plant colonization genes.
PLoS biology (2017-09-25)
Benjamin J Cole, Meghan E Feltcher, Robert J Waters, Kelly M Wetmore, Tatiana S Mucyn, Elizabeth M Ryan, Gaoyan Wang, Sabah Ul-Hasan, Meredith McDonald, Yasuo Yoshikuni, Rex R Malmstrom, Adam M Deutschbauer, Jeffery L Dangl, Axel Visel
ABSTRACT
Diverse soil-resident bacteria can contribute to plant growth and health, but the molecular mechanisms enabling them to effectively colonize their plant hosts remain poorly understood. We used randomly barcoded transposon mutagenesis sequencing (RB-TnSeq) in Pseudomonas simiae, a model root-colonizing bacterium, to establish a genome-wide map of bacterial genes required for colonization of the Arabidopsis thaliana root system. We identified 115 genes (2% of all P. simiae genes) with functions that are required for maximal competitive colonization of the root system. Among the genes we identified were some with obvious colonization-related roles in motility and carbon metabolism, as well as 44 other genes that had no or vague functional predictions. Independent validation assays of individual genes confirmed colonization functions for 20 of 22 (91%) cases tested. To further characterize genes identified by our screen, we compared the functional contributions of P. simiae genes to growth in 90 distinct in vitro conditions by RB-TnSeq, highlighting specific metabolic functions associated with root colonization genes. Our analysis of bacterial genes by sequence-driven saturation mutagenesis revealed a genome-wide map of the genetic determinants of plant root colonization and offers a starting point for targeted improvement of the colonization capabilities of plant-beneficial microbes.
Sigma-Aldrich
Membrane Scaffold Protein 1D1, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Phytagel, BioReagent, suitable for plant cell culture, powder