MilliporeSigma
  • RHAMM expression and isoform balance predict aggressive disease and poor survival in multiple myeloma.
RHAMM expression and isoform balance predict aggressive disease and poor survival in multiple myeloma.
Blood (2004-04-24)
Christopher A Maxwell, Erik Rasmussen, Fenghuang Zhan, Jonathan J Keats, Sophia Adamia, Erin Strachan, Mary Crainie, Ronald Walker, Andrew R Belch, Linda M Pilarski, Bart Barlogie, John Shaughnessy, Tony Reiman
ABSTRACT
Multiple myeloma (MM) plasma cells (PCs) express receptor for hyaluronan-mediated motility (RHAMM), a hyaluronan-binding, cytoskeleton and centrosome protein. The most abundant RHAMM isoforms in MM are full-length RHAMM (RHAMMFL) and the splice variant RHAMM-exon4. We separately examined the significance of RHAMM expression, and isoform balance, in 2 groups of MM patients. In oligonucleotide microarray experiments (n=210, Arkansas), increasing RHAMM mRNA expression in MM PCs is strongly associated with osteolytic bone lesions (P <.001), and event-free (P =.05) and overall (P =.04) survival. Semiquantitative determination of RHAMM isoform expression (Alberta, Canada) used capillary electrophoretic detection and measurement of RHAMM-exon4/RHAMMFL reverse-transcriptase-polymerase chain reaction (RT-PCR) products. RHAMM isoforms are rarely expressed concurrently in single MM PCs; the pattern of isoform expression, at the single-cell level, is approximated in larger numbers of cells by the RHAMM-exon4/RHAMMFL ratio. Absolute RHAMM expression and the RHAMM-exon4/RHAMMFL ratio are only partially correlated in MM PCs; in cell lines, absolute RHAMM expression is elevated in mitosis, while RHAMM ratios remain stable. Temporal examination of MM patients' peripheral blood reveals that the RHAMM-exon4/RHAMMFL ratio increases with disease burden. The RHAMM-exon4/RHAMMFL ratio in diagnostic bone marrow samples (n=101, Alberta) is an independent prognostic factor. Thus, expression and splicing of RHAMM are important molecular determinants of disease severity in MM.
Sigma-Aldrich
Vitamins Kit, ~98% (Components, TLC)
Pricing and availability is not currently available.