MilliporeSigma
  • Rationally designed BCL6 inhibitors target activated B cell diffuse large B cell lymphoma.
Rationally designed BCL6 inhibitors target activated B cell diffuse large B cell lymphoma.
The Journal of clinical investigation (2016-08-03)
Mariano G Cardenas, Wenbo Yu, Wendy Beguelin, Matthew R Teater, Huimin Geng, Rebecca L Goldstein, Erin Oswald, Katerina Hatzi, Shao-Ning Yang, Joanna Cohen, Rita Shaknovich, Kenno Vanommeslaeghe, Huimin Cheng, Dongdong Liang, Hyo Je Cho, Joshua Abbott, Wayne Tam, Wei Du, John P Leonard, Olivier Elemento, Leandro Cerchietti, Tomasz Cierpicki, Fengtian Xue, Alexander D MacKerell, Ari M Melnick
ABSTRACT
Diffuse large B cell lymphomas (DLBCLs) arise from proliferating B cells transiting different stages of the germinal center reaction. In activated B cell DLBCLs (ABC-DLBCLs), a class of DLBCLs that respond poorly to current therapies, chromosomal translocations and amplification lead to constitutive expression of the B cell lymphoma 6 (BCL6) oncogene. The role of BCL6 in maintaining these lymphomas has not been investigated. Here, we designed small-molecule inhibitors that display higher affinity for BCL6 than its endogenous corepressor ligands to evaluate their therapeutic efficacy for targeting ABC-DLBCL. We used an in silico drug design functional-group mapping approach called SILCS to create a specific BCL6 inhibitor called FX1 that has 10-fold greater potency than endogenous corepressors and binds an essential region of the BCL6 lateral groove. FX1 disrupted formation of the BCL6 repression complex, reactivated BCL6 target genes, and mimicked the phenotype of mice engineered to express BCL6 with corepressor binding site mutations. Low doses of FX1 induced regression of established tumors in mice bearing DLBCL xenografts. Furthermore, FX1 suppressed ABC-DLBCL cells in vitro and in vivo, as well as primary human ABC-DLBCL specimens ex vivo. These findings indicate that ABC-DLBCL is a BCL6-dependent disease that can be targeted by rationally designed inhibitors that exceed the binding affinity of natural BCL6 ligands.
Sigma-Aldrich
FX1, ≥98% (HPLC)
Pricing and availability is not currently available.