MilliporeSigma
  • Distribution and Molecular Characterization of Campylobacter Species at Different Processing Stages in Two Poultry Processing Plants.
Distribution and Molecular Characterization of Campylobacter Species at Different Processing Stages in Two Poultry Processing Plants.
Foodborne pathogens and disease (2017-02-06)
Soo-Kyoung Lee, Hyun-Jung Park, Jin-Hee Lee, Jong-Soo Lim, Kun-Ho Seo, Eun-Jeong Heo, Young-Jo Kim, Sung-Hwan Wee, Jin-San Moon
ABSTRACT
The present study analyzed the prevalence and molecular characterization of Campylobacter at different processing steps in poultry slaughterhouses to determine where contamination mainly occurs. A total of 1,040 samples were collected at four different stages (preprocessing cloacal swabs, postevisceration, postwashing, and postchilling) in two processing plants. Campylobacter was detected in 5.8% (15 of 260) of the cloacal swabs and in 13.3% (104 of 780) of the processing samples. In both plants, the sampling points with the greatest contamination rates were after evisceration (20.5% and 15.4% for plants A and B, respectively) and significantly decreased after chilling (p < 0.05, from 20.5% to 10.9%) in plant A and after washing (from 15.4% to 2.9%) in plants B. In the result, however, the reduction in Campylobacter contamination was achieved through the sequential processing procedures in both plants. Campylobacter loads (>10
Sigma-Aldrich
Indoxyl acetate, ≥95% (GC)