MilliporeSigma
  • Remobilization of polycyclic aromatic hydrocarbons and organic matter in seawater during sediment resuspension experiments from a polluted coastal environment: Insights from Toulon Bay (France).
Remobilization of polycyclic aromatic hydrocarbons and organic matter in seawater during sediment resuspension experiments from a polluted coastal environment: Insights from Toulon Bay (France).
Environmental pollution (Barking, Essex : 1987) (2017-07-10)
Catherine Guigue, Marc Tedetti, Duc Huy Dang, Jean-Ulrich Mullot, Cédric Garnier, Madeleine Goutx
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs) and organic matter contents were measured in seawater during resuspension experiments using sediments collected from Toulon Bay (Northwestern Mediterranean Sea, France). The studied sediments were very highly contaminated in PAHs, especially in 4-ring compounds emitted from combustion processes. The sediments used for resuspension experiments were collected at 0-2 cm (diagenetically new organic matter, OM) and 30-32 cm depths (diagenetically transformed OM). They were both mostly composed of fine particles (<63 μm), enriched in organic carbon (8.2 and 6.3%, respectively) and in PAHs (concentration of Σ34 PAHs: 38.2 and 35.7 × 10
Product Description
Supelco
Alkane standard solution C21-C40, analytical standard