MilliporeSigma
  • Increased circular RNA UBAP2 acts as a sponge of miR-143 to promote osteosarcoma progression.
Increased circular RNA UBAP2 acts as a sponge of miR-143 to promote osteosarcoma progression.
Oncotarget (2017-10-06)
Hao Zhang, Guangchao Wang, Chen Ding, Peng Liu, Renkai Wang, Wenbin Ding, Dake Tong, Dajiang Wu, Cheng Li, Qiang Wei, Xin Zhang, Di Li, Peizhao Liu, Haochen Cui, Hao Tang, Fang Ji
ABSTRACT
Deregulated expression of circular RNA (circRNA) has been determined to be important in carcinogenesis and progression; however, in the most common type of primary malignant bone tumor osteosarcoma, the roles of circRNA in cancer development still remain to be elucidated. Here, we found that circRNA UBAP2 (circUBAP2) expression is significantly increased in human osteosarcoma tissues as compared to those in matched controls. Increased circUBAP2 expression was significantly correlated with human osteosarcoma progression and prognosis. Furthermore, increased circUBAP2 could promote osteosarcoma growth and inhibit apoptosis both
Sigma-Aldrich
MISSION® esiRNA, targeting human UBAP2
Pricing and availability is not currently available.