MilliporeSigma
  • Enhanced bone formation in the vicinity of porous β-TCP scaffolds exhibiting slow release of collagen-derived tripeptides.
Enhanced bone formation in the vicinity of porous β-TCP scaffolds exhibiting slow release of collagen-derived tripeptides.
Journal of materials science. Materials in medicine (2017-07-27)
Keita Kamikura, Tsutomu Minatoya, Michiko Terada-Nakaishi, Shoko Yamamoto, Yasuo Sakai, Toshitake Furusawa, Yuta Matsushima, Hidero Unuma
ABSTRACT
It has been experimentally proven that orally ingested collagen-derived tripeptides (Ctp) are quickly absorbed in the body and effectively promote the regeneration of connective tissues including bone and skin. Ctp are capable to activate osteoblasts and fibroblasts, which eventually promotes tissue regeneration. Based on these findings, a hypothesis was formulated in this study that direct delivery of Ctp to bone defect would also facilitate tissue regeneration as well as oral administration. To test the hypothesis, we prepared a bone augmentation material with the ability to slowly release Ctp, and investigated its in vivo bone regeneration efficacy. The implant material was porous β-tricalcium phosphate (β-TCP) scaffold which was coated with a co-precipitated layer of bone-like hydroxyapatite and Ctp. The β-TCP was impregnated with approximately 0.8%(w/w) Ctp. Then, the Ctp-modified β-TCP was implanted into bone defects of Wistar rats to evaluate in vivo efficacy of Ctp directly delivered from the material to the bone defects. The control was pristine porous β-TCP. In vitro tests showed that Ctp were steadily released from the co-precipitated layer for approximately two weeks. The Ctp-modified scaffolds significantly promoted new bone formation in vivo in their vicinity as compared with pristine β-TCP scaffolds; 6 weeks after the implantation, Ctp-modified scaffolds promoted twice as much bone formation as the control implants. Consequently, we achieved the slow and steady release of Ctp, and found that direct delivery of Ctp from implant materials was effective for bone regeneration as well as oral administration. A β-TCP scaffold capable of slowly releasing bone-enhancing substances significantly promoted bone formation.
Sigma-Aldrich
Cefazolin sodium salt, 89.1-110.1%