MilliporeSigma
Fabrication of three-dimensional ordered macroporous spinel CoFe
Nanoscale (2017-04-08)
Jong Guk Kim, Yuseong Noh, Youngmin Kim, Seonhwa Lee, Won Bae Kim
ABSTRACT
Three-dimensionally ordered macroporous (3DOM) CoFe
Sigma-Aldrich
Nitrogen-14N2, 99.99 atom % 14N