MilliporeSigma
  • CHASE-Containing Histidine Kinase Receptors in Apple Tree: From a Common Receptor Structure to Divergent Cytokinin Binding Properties and Specific Functions.
CHASE-Containing Histidine Kinase Receptors in Apple Tree: From a Common Receptor Structure to Divergent Cytokinin Binding Properties and Specific Functions.
Frontiers in plant science (2017-10-06)
Dimitri Daudu, Elsa Allion, Franziska Liesecke, Nicolas Papon, Vincent Courdavault, Thomas Dugé de Bernonville, Céline Mélin, Audrey Oudin, Marc Clastre, Arnaud Lanoue, Martine Courtois, Olivier Pichon, David Giron, Sabine Carpin, Nathalie Giglioli-Guivarc'h, Joël Crèche, Sébastien Besseau, Gaëlle Glévarec
ABSTRACT
Cytokinin signaling is a key regulatory pathway of many aspects in plant development and environmental stresses. Herein, we initiated the identification and functional characterization of the five CHASE-containing histidine kinases (CHK) in the economically important
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
L-(−)-Galactose, ≥99% (HPLC)