MilliporeSigma
  • Block copolymer-boron cluster conjugate for effective boron neutron capture therapy of solid tumors.
Block copolymer-boron cluster conjugate for effective boron neutron capture therapy of solid tumors.
Journal of controlled release : official journal of the Controlled Release Society (2017-03-25)
Peng Mi, Hironobu Yanagie, Novriana Dewi, Hung-Chi Yen, Xueying Liu, Minoru Suzuki, Yoshinori Sakurai, Koji Ono, Hiroyuki Takahashi, Horacio Cabral, Kazunori Kataoka, Nobuhiro Nishiyama
ABSTRACT
Boron neutron capture therapy is a promising tumor treatment method, though its wide application has been limited due to the poor tumor selectivity and intracellular delivery of
Sigma-Aldrich
Benzenesulfonyl hydrazide, 98%