MilliporeSigma
  • Enhanced insulin secretion and insulin sensitivity in young lambs with placental insufficiency-induced intrauterine growth restriction.
Enhanced insulin secretion and insulin sensitivity in young lambs with placental insufficiency-induced intrauterine growth restriction.
American journal of physiology. Regulatory, integrative and comparative physiology (2017-05-12)
Leticia E Camacho, Xiaochuan Chen, William W Hay, Sean W Limesand
ABSTRACT
Intrauterine growth restriction (IUGR) is associated with persistent metabolic complications, but information is limited for IUGR infants. We determined glucose-stimulated insulin secretion (GSIS) and insulin sensitivity in young lambs with placental insufficiency-induced IUGR. Lambs with hyperthermia-induced IUGR (
Sigma-Aldrich
CelLytic MT Cell Lysis Reagent, For mammalian tissues