MilliporeSigma
  • Comparison of cerato-platanin family protein BcSpl1 produced in Pichia pastoris and Escherichia coli.
Comparison of cerato-platanin family protein BcSpl1 produced in Pichia pastoris and Escherichia coli.
Protein expression and purification (2017-06-14)
Yi Zhang, Yingbo Liang, Dewen Qiu, Jingjing Yuan, Xiufen Yang
ABSTRACT
The Botrytis cinerea BcSpl1 protein is a member of the cerato-platanin family, and consists of 137 amino acids and two disulfide bridges. This protein induces the onset of necrosis in infiltrated plant hosts. Recombinant BcSpl1 proteins produced in Pichia pastoris (pBcSpl1) and Escherichia coli (eBcSpl1) were initially compared regarding their abilities to induce necrosis and systemic acquired resistance (SAR). The pBcSpl1 and eBcSpl1 treatments led to the development of necrotic lesions on tomato leaves, and provided tomato plants with SAR to B. cinerea. The lesion area of leaves infiltrated with the BcSpl1 proteins decreased by 22.7% (pBcSpl1) and 21.8% (eBcSpl1). Additionally, eBcSpl1 up-regulated the expression levels of some defense-related genes, including PR-1a, prosystemin, PI I, and PI II, as well as SIPK and TPK1b, which encode two protein kinases. Furthermore, eBcSpl1 exhibited chitin-binding properties. Our data revealed that the E. coli expression system produces higher BcSpl1 yields than the P. pastoris system. This high-yield expression of BcSpl1 may be relevant for future large-scale applications of this elicitor to improve crop production.
Sigma-Aldrich
Chitin from shrimp shells, suitable for analysis of chitinase, BioReagent, purified powder
Pricing and availability is not currently available.