MilliporeSigma
Tunable Length and Optical Properties of CsPbX
Nano letters (2017-01-18)
Daniel Amgar, Avigail Stern, Dvir Rotem, Danny Porath, Lioz Etgar
ABSTRACT
Perovskite nanostructures, both hybrid organo-metal and fully inorganic perovskites, have gained a lot of interest in the past few years for their intriguing optical properties in the visible region. We report on inorganic cesium lead bromide (CsPbBr
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Pricing and availability is not currently available.