- 4-1BB-Enhanced Expansion of CD8
4-1BB-Enhanced Expansion of CD8
Cancer immunology research (2017-05-06)
Michiko Harao, Marie-Andrée Forget, Jason Roszik, Hui Gao, Gildy V Babiera, Savitri Krishnamurthy, Jessica A Chacon, Shumin Li, Elizabeth A Mittendorf, Sarah M DeSnyder, Korrene F Rockwood, Chantale Bernatchez, Naoto T Ueno, Laszlo G Radvanyi, Luis Vence, Cara Haymaker, James M Reuben
PMID28473315
ABSTRACT
Triple-negative breast cancer (TNBC) highly infiltrated with CD8